Anti-APOE Mouse Monoclonal Antibody [clone: 1H4]

Lieferant: Abnova
ABNOMAB10490EA 518 EUR
ABNOMAB10490
Anti-APOE Mouse Monoclonal Antibody [clone: 1H4]
Antikörper
Mouse monoclonal antibody raised against partial recombinant APOE.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: APOE
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR