Anti-ANIB1 Mouse Monoclonal Antibody [clone: 5H3]

Lieferant: Abnova
ABNOMAB10483EA 454 EUR
ABNOMAB10483
Anti-ANIB1 Mouse Monoclonal Antibody [clone: 5H3]
Antikörper
Mouse monoclonal antibody raised against partial recombinant ANIB1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: ANIB1
Clonality: Monoclonal
Clone: 5H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR