Anti-TYRO3 Mouse Monoclonal Antibody [clone: 10E11]

Lieferant: Abnova
ABNOMAB10475EA 370 EUR
ABNOMAB10475
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 10E11]
Antikörper
Mouse monoclonal antibody raised against partial recombinant TYRO3.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 10E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR