Anti-IgG Mouse Monoclonal Antibody [clone: 4A10]

Lieferant: Abnova
ABNOMAB10474EA 370 EUR
ABNOMAB10474
Anti-IgG Mouse Monoclonal Antibody [clone: 4A10]
Antikörper
Mouse monoclonal antibody raised against recombinant IgG (Fc).

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; ELISA: 1:10001

Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone: 4A10
Conjugation: Unconjugated
Epitope: F(c)
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR