Anti-LOC541473 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00541473-D01PEA 368 EUR
ABNOH00541473-D01P
Anti-LOC541473 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human LOC541473 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: LOC541473
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR