Anti-EV71 VP1 Mouse Monoclonal Antibody [clone: 3D7]

Lieferant: Abnova
ABNOMAB1255-M05EA 382 EUR
ABNOMAB1255-M05
Anti-EV71 VP1 Mouse Monoclonal Antibody [clone: 3D7]
Antikörper
Mouse monoclonal antibody raised against a full length recombinant virus EV71 VP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: EV71 VP1
Clonality: Monoclonal
Clone: 3D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR