Anti-KDM5C Mouse Monoclonal Antibody

Lieferant: Abnova
ABNOMAB11151EA 350 EUR
ABNOMAB11151
Anti-KDM5C Mouse Monoclonal Antibody
Antikörper
Mouse monoclonal antibody raised against partial recombinant protein of JARID1C.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:100-1:2000

Type: Primary
Antigen: KDM5C
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR