Anti-ABCC11 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24793EA 454 EUR
ABNOPAB24793
Anti-ABCC11 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ABCC11.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ABCC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR