Anti-Cetuximab Monoclonal Antibody [clone: 6D4-9]

Lieferant: Abnova
ABNOMAB11129EA 684 EUR
ABNOMAB11129
Anti-Cetuximab Monoclonal Antibody [clone: 6D4-9]
Antikörper
Mouse monoclonal antibody raised against F(ab)2 fragment of Cetuximab.

Recommended Dilutions: ELISA: 2 ?g/ml for coating

Type: Primary
Antigen: Cetuximab
Clonality: Monoclonal
Clone: 6D4-9
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR