Anti-Bevacizumab Monoclonal Antibody [clone: 2C8-13]

Lieferant: Abnova
ABNOMAB11128EA 772 EUR
ABNOMAB11128
Anti-Bevacizumab Monoclonal Antibody [clone: 2C8-13]
Antikörper
Mouse monoclonal antibody raised against F(ab)2 fragment of Bevacizumab.

Recommended Dilutions: ELISA: 2 ?g/ml for coating

Type: Primary
Antigen: Bevacizumab
Clonality: Monoclonal
Clone: 2C8-13
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR