Anti-ACTB Mouse Monoclonal Antibody [clone: M01]

Lieferant: Abnova
ABNOMAB11127EA 568 EUR
ABNOMAB11127
Anti-ACTB Mouse Monoclonal Antibody [clone: M01]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of ACTB.

Recommended Dilutions: Western Blot: 1:2000-1:5000

Type: Primary
Antigen: ACTB
Clonality: Monoclonal
Clone: M01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR