Anti-MITF Mouse Monoclonal Antibody [clone: D5]

Lieferant: Abnova
ABNOMAB11118EA 793 EUR
ABNOMAB11118
Anti-MITF Mouse Monoclonal Antibody [clone: D5]
Antikörper
Mouse monoclonal antibody raised against recombinant protein of human MITF.

Type: Primary
Antigen: MITF
Clonality: Monoclonal
Clone: D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR