Anti-PKLR Mouse Monoclonal Antibody [clone: AT1E3]

Lieferant: Abnova
ABNOMAB11108EA 350 EUR
ABNOMAB11108
Anti-PKLR Mouse Monoclonal Antibody [clone: AT1E3]
Antikörper
Mouse monoclonal antibody raised against partial recombinant PKLR.

Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: PKLR
Clonality: Monoclonal
Clone: AT1E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR