Anti-GAPDH Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24753EA 454 EUR
ABNOPAB24753
Anti-GAPDH Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of GAPDH.

Recommended Dilutions: Western Blot: 1:5000-1:10000

Type: Primary
Antigen: GAPDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR