Anti-TUBB Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24751EA 518 EUR
ABNOPAB24751
Anti-TUBB Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of TUBB 1/2/3/5/6/8.

Recommended Dilutions: Western Blot: 1:5000-1:10000

Type: Primary
Antigen: TUBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR