Anti-ACTB Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24749EA 454 EUR
ABNOPAB24749
Anti-ACTB Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ACTB.

  • Recommended Dilutions: Western Blot: 1:3000-1:10000

Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR