Anti-SOX2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24743EA 454 EUR
ABNOPAB24743
Anti-SOX2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SOX2.

Recommended Dilutions: Western Blot: 1:1000-1:5000

Type: Primary
Antigen: SOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR