Anti-APP Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24741EA 454 EUR
ABNOPAB24741
Anti-APP Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of Amyloid beta 1-16.

  • Recommended Dilutions: Western Blot: 1:1000-1:5000

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR