Anti-STAT1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24731EA 568 EUR
ABNOPAB24731
Anti-STAT1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Stat1.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR