Anti-CARD17 Mouse Monoclonal Antibody [clone: 1G6]

Lieferant: Abnova
ABNOH00440068-M01AEA 382 EUR
ABNOH00440068-M01A
Anti-CARD17 Mouse Monoclonal Antibody [clone: 1G6]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant CARD17.

Type: Primary
Antigen: CARD17
Clonality: Monoclonal
Clone: 1G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR