Anti-SLC9A3 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24716EA 454 EUR
ABNOPAB24716
Anti-SLC9A3 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SLC9A3.

Recommended Dilutions: ELISA: 1:10000; Dot Blot: 1:10000; Western Blot: 1:500

Type: Primary
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR