Anti-RCAN1 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24686EA 370 EUR
ABNOPAB24686
Anti-RCAN1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of RCAN1.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: RCAN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR