Anti-S100 A8 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25443EA 370 EUR
ABNOPAB25443
Anti-S100 A8 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of S100A8.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.03-0.1 ?g/ml

Type: Primary
Antigen: S100 A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR