Anti-MAP2K4 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25412EA 419 EUR
ABNOPAB25412
Anti-MAP2K4 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP2K4.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR