Anti-STMN1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25371EA 419 EUR
ABNOPAB25371
Anti-STMN1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of STMN1.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: STMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR