Anti-PTK2B Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25352EA 370 EUR
ABNOPAB25352
Anti-PTK2B Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PTK2B.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: PTK2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR