Anti-ZAP70 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24606EA 518 EUR
ABNOPAB24606
Anti-ZAP70 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of ZAP70.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: ZAP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR