Anti-MTHFR Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24568EA 370 EUR
ABNOPAB24568
Anti-MTHFR Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of MTHFR.

Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: MTHFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR