Anti-KCNC3 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24567EA 370 EUR
ABNOPAB24567
Anti-KCNC3 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of KCNC3.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: KCNC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR