Anti-SFTPA1 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24551EA 518 EUR
ABNOPAB24551
Anti-SFTPA1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of SFTPA1/SFTPA2.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.01-0.03 ?g/ml

Type: Primary
Antigen: SFTPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR