Anti-LYZ Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB24550EA 568 EUR
ABNOPAB24550
Anti-LYZ Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against full length native LYZ.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:500-1:2000; Immunoprecipitation: 1:100

Type: Primary
Antigen: LYZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR