Anti-MEF2A Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25261EA 370 EUR
ABNOPAB25261
Anti-MEF2A Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MEF2A.

Recommended Dilutions: Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: MEF2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR