Anti-ELK1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25258EA 370 EUR
ABNOPAB25258
Anti-ELK1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ELK1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: ELK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR