Anti-DSG4 Mouse Monoclonal Antibody [clone: 6E7]

Lieferant: Abnova
ABNOH00147409-M06EA 382 EUR
ABNOH00147409-M06
Anti-DSG4 Mouse Monoclonal Antibody [clone: 6E7]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant DSG4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: DSG4
Clonality: Monoclonal
Clone: 6E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR