Anti-BORCS5 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00118426-B01PEA 394 EUR
ABNOH00118426-B01P
Anti-BORCS5 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human LOH12CR1 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: BORCS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human