Anti-MYLK4 Mouse Monoclonal Antibody [clone: 7A3]

Lieferant: Abnova
ABNOH00340156-M01EA 382 EUR
ABNOH00340156-M01
Anti-MYLK4 Mouse Monoclonal Antibody [clone: 7A3]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant MYLK4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: MYLK4
Clonality: Monoclonal
Clone: 7A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR