Anti-ARL13B Mouse Monoclonal Antibody [clone: 7G9]

Lieferant: Abnova
ABNOH00200894-M01AEA 394 EUR
ABNOH00200894-M01A
Anti-ARL13B Mouse Monoclonal Antibody [clone: 7G9]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant ARL13B.

Type: Primary
Antigen: ARL13B
Clonality: Monoclonal
Clone: 7G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR