Anti-FOXD4L1 Mouse Monoclonal Antibody [clone: 8A8]

Lieferant: Abnova
ABNOH00200350-M04EA 382 EUR
ABNOH00200350-M04
Anti-FOXD4L1 Mouse Monoclonal Antibody [clone: 8A8]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant FOXD4L1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: FOXD4L1
Clonality: Monoclonal
Clone: 8A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR