Anti-OSBPL7 Mouse Monoclonal Antibody [clone: 3D11]

Lieferant: Abnova
ABNOH00114881-M03EA 382 EUR
ABNOH00114881-M03
Anti-OSBPL7 Mouse Monoclonal Antibody [clone: 3D11]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant OSBPL7.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: OSBPL7
Clonality: Monoclonal
Clone: 3D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR