Anti-C1QL3 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00389941-B01PEA 394 EUR
ABNOH00389941-B01P
Anti-C1QL3 Mouse Polyclonal Antibody
Antikörper
Mouse polyclonal antibody raised against a full-length human C1QL3 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: C1QL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human