Anti-TMEM86B Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00255043-D01EA 380 EUR
ABNOH00255043-D01
Anti-TMEM86B Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against a full-length human TMEM86B protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TMEM86B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR