Anti-NFAM1 Mouse Monoclonal Antibody [clone: 2E9]
Lieferant: Abnova
ABNOH00150372-M10EA 394 EUR
ABNOH00150372-M10
Anti-NFAM1 Mouse Monoclonal Antibody [clone: 2E9]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant NFAM1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: NFAM1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human