Anti-LRRN1 Mouse Monoclonal Antibody [clone: 4G8]
Lieferant: Abnova
ABNOH00057633-M09EA 394 EUR
ABNOH00057633-M09
Anti-LRRN1 Mouse Monoclonal Antibody [clone: 4G8]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant LRRN1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: LRRN1
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human