Anti-EBF4 Mouse Monoclonal Antibody [clone: 4E10]

Lieferant: Abnova
ABNOH00057593-M04EA 382 EUR
ABNOH00057593-M04
Anti-EBF4 Mouse Monoclonal Antibody [clone: 4E10]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant EBF4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: EBF4
Clonality: Monoclonal
Clone: 4E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR