Anti-PDCD1LG2 Mouse Monoclonal Antibody [clone: 7D5]

Lieferant: Abnova
ABNOH00080380-M06EA 382 EUR
ABNOH00080380-M06
Anti-PDCD1LG2 Mouse Monoclonal Antibody [clone: 7D5]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant PDCD1LG2.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PDCD1LG2
Clonality: Monoclonal
Clone: 7D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR