Anti-SLC30A5 Mouse Monoclonal Antibody [clone: 2E10]

Lieferant: Abnova
ABNOH00064924-M03AEA 382 EUR
ABNOH00064924-M03A
Anti-SLC30A5 Mouse Monoclonal Antibody [clone: 2E10]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant SLC30A5.

Type: Primary
Antigen: SLC30A5
Clonality: Monoclonal
Clone: 2E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR