Anti-ELOF1 Mouse Monoclonal Antibody [clone: 4F6]
Lieferant: Abnova
ABNOH00084337-M06EA 394 EUR
ABNOH00084337-M06
Anti-ELOF1 Mouse Monoclonal Antibody [clone: 4F6]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant ELOF1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: ELOF1
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human