Anti-EXOSC5 Mouse Monoclonal Antibody [clone: 2E7]

Lieferant: Abnova
ABNOH00056915-M03EA 382 EUR
ABNOH00056915-M03
Anti-EXOSC5 Mouse Monoclonal Antibody [clone: 2E7]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant EXOSC5.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: EXOSC5
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR