Anti-BAIAP2L1 Mouse Monoclonal Antibody [clone: 4A7]

Lieferant: Abnova
ABNOH00055971-M09EA 382 EUR
ABNOH00055971-M09
Anti-BAIAP2L1 Mouse Monoclonal Antibody [clone: 4A7]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant BAIAP2L1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: BAIAP2L1
Clonality: Monoclonal
Clone: 4A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR