Anti-MID1IP1 Mouse Monoclonal Antibody [clone: 8G8]

Lieferant: Abnova
ABNOH00058526-M01EA 382 EUR
ABNOH00058526-M01
Anti-MID1IP1 Mouse Monoclonal Antibody [clone: 8G8]
Antikörper
Mouse monoclonal antibody raised against a full-length recombinant MID1IP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: MID1IP1
Clonality: Monoclonal
Clone: 8G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR